missing translation for 'onlineSavingsMsg'
Learn More

9(10H)-Acridone, 99%

Catalog No. p-50508
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC400250100 10 g Glass bottle
AC400250250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC400250100 Supplier Thermo Scientific Chemicals Supplier No. 400250100
Only null left

CAS: 578-95-0 | C13H9NO | 195.22 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 578-95-0
Molecular Formula C13H9NO
Molecular Weight (g/mol) 195.22
MDL Number MFCD00005019
InChI Key FZEYVTFCMJSGMP-UHFFFAOYSA-N
Synonym acridone, 9 10h-acridone, acridin-9 10h-one, 9-acridone, 9 10h-acridinone, acridanone, 9-acridanone, acridin-9-one, 9-acridinol, 9,10-dihydro-9-oxoacridine
PubChem CID 2015
ChEBI CHEBI:50756
IUPAC Name 10H-acridin-9-one
SMILES C1=CC=C2C(=C1)C(=O)C3=CC=CC=C3N2

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 360.0°C to 362.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 10 g
Formula Weight 195.22
Percent Purity 99%
Physical Form Crystalline Powder
Chemical Name or Material 9(10H)-Acridone, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 209-434-7
RTECSNumber AR6976000
TSCA TSCA

RUO – Research Use Only