missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydroacridine 98.0+%, TCI America™

Catalog No. p-7128634
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D50581G 1 g
D50585G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D50581G Supplier TCI America Supplier No. D50581G
Only null left

Chemical Identifiers

CAS 92-81-9
Molecular Formula C13H11N
Molecular Weight (g/mol) 181.238
MDL Number MFCD00022256
InChI Key HJCUTNIGJHJGCF-UHFFFAOYSA-N
PubChem CID 7106
IUPAC Name 9,10-dihydroacridine
SMILES C1C2=CC=CC=C2NC3=CC=CC=C31

Specifications

Melting Point 171°C
Color White-Yellow
Boiling Point 348°C
Quantity 1 g
Formula Weight 181.24
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydroacridine
RTECSNumber AR6950000
TSCA No