missing translation for 'onlineSavingsMsg'
Learn More

9,10-Dihydro-9-oxa-10-phosphaphenanthrene 10-Oxide 97.0+%, TCI America™

Catalog No. p-7126985
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D187425G 25 g
D1874100G 100 g
D1874500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D187425G Supplier TCI America Supplier No. D187425G
Only null left

Chemical Identifiers

CAS 35948-25-5
Molecular Formula C12H8O2P
Molecular Weight (g/mol) 215.17
MDL Number MFCD00040561
InChI Key DWSWCPPGLRSPIT-UHFFFAOYNA-N
Synonym 3,4:5,6-Dibenzo-1,2-oxaphosphane-2-oxide
PubChem CID 6328250
IUPAC Name 9-oxo-8-oxa-9λ⁵-phosphatricyclo[8.4.0.0²,⁷]tetradeca-1(14),2,4,6,10,12-hexaen-9-ylium
SMILES O=[P+]1OC2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 119°C
Color White
Quantity 25 g
Formula Weight 216.18
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Dihydro-9-oxa-10-phosphaphenanthrene 10-Oxide
EINECSNumber (5)-3777
TSCA Yes