missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Dihydro-9,9-dimethylacridine 98.0+%, TCI America™
$119.30 - $1066.24
Chemical Identifiers
| CAS | 6267-02-3 |
|---|---|
| Molecular Formula | C15H15N |
| Molecular Weight (g/mol) | 209.29 |
| MDL Number | MFCD00030130 |
| InChI Key | JSEQNGYLWKBMJI-UHFFFAOYSA-N |
| Synonym | 9,9-Dimethyl-9,10-dihydroacridine |
| PubChem CID | 22647 |
| IUPAC Name | 9,9-dimethyl-10H-acridine |
| SMILES | CC1(C2=CC=CC=C2NC3=CC=CC=C31)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
D5028200MG
|
TCI America
D5028200MG |
200 mg |
Each for $119.30
|
|
|||||
|
D50281G
|
TCI America
D50281G |
1 g |
Each for $354.17
|
|
|||||
|
D50285G
|
TCI America
D50285G |
5 g |
Each for $1,066.24
|
|
|||||
Chemical Identifiers
| 6267-02-3 | |
| 209.29 | |
| JSEQNGYLWKBMJI-UHFFFAOYSA-N | |
| 22647 | |
| CC1(C2=CC=CC=C2NC3=CC=CC=C31)C |
| C15H15N | |
| MFCD00030130 | |
| 9,9-Dimethyl-9,10-dihydroacridine | |
| 9,9-dimethyl-10H-acridine |
Specifications
| 6267-02-3 | |
| White-Yellow | |
| C15H15N | |
| 200 mg | |
| JSEQNGYLWKBMJI-UHFFFAOYSA-N | |
| 9,9-dimethyl-10H-acridine | |
| 22647 | |
| ≥98.0% (GC) | |
| 9,10-Dihydro-9,9-dimethylacridine |
| 127°C | |
| 155°C | |
| MFCD00030130 | |
| 9,9-Dimethyl-9,10-dihydroacridine | |
| CC1(C2=CC=CC=C2NC3=CC=CC=C31)C | |
| 209.29 | |
| 209.29 | |
| Crystalline Powder |
Safety and Handling
TSCA : Yes