missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Dihydro-9,9-dimethylacridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 127°C |
| Color | White-Yellow |
| Boiling Point | 155°C |
| Quantity | 200 mg |
| Formula Weight | 209.29 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9,10-Dihydro-9,9-dimethylacridine |
Safety and Handling
| TSCA | Yes |