missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Bis[N-(m-tolyl)anilino]anthracene 98.0+%, TCI America™
$130.48 - $449.24
Chemical Identifiers
| CAS | 189263-81-8 |
|---|---|
| Molecular Formula | C40H32N2 |
| Molecular Weight (g/mol) | 540.71 |
| InChI Key | LMTXHBDTVAIIQY-UHFFFAOYSA-N |
| Synonym | N,N′C-Diphenyl-N,N′C-di(m-tolyl)-9,10-anthracenediamine, N,N′C-Bis(3-methylphenyl)-N,N′C-diphenyl-9,10-anthracenediamine |
| PubChem CID | 22976649 |
| IUPAC Name | 9-N,10-N-bis(3-methylphenyl)-9-N,10-N-diphenylanthracene-9,10-diamine |
| SMILES | CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=CC=C6)C7=CC=CC(=C7)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4965200MG
|
TCI America
B4965200MG |
200 mg |
Each for $130.48
|
|
|||||
|
B49651G
|
TCI America
B49651G |
1 g |
Each for $449.24
|
|
|||||
Chemical Identifiers
| 189263-81-8 | |
| 540.71 | |
| N,N′C-Diphenyl-N,N′C-di(m-tolyl)-9,10-anthracenediamine, N,N′C-Bis(3-methylphenyl)-N,N′C-diphenyl-9,10-anthracenediamine | |
| 9-N,10-N-bis(3-methylphenyl)-9-N,10-N-diphenylanthracene-9,10-diamine |
| C40H32N2 | |
| LMTXHBDTVAIIQY-UHFFFAOYSA-N | |
| 22976649 | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=CC=C6)C7=CC=CC(=C7)C |
Specifications
| 189263-81-8 | |
| Yellow | |
| 200 mg | |
| LMTXHBDTVAIIQY-UHFFFAOYSA-N | |
| 9-N,10-N-bis(3-methylphenyl)-9-N,10-N-diphenylanthracene-9,10-diamine | |
| 22976649 | |
| ≥98.0% (HPLC) | |
| 9,10-Bis[N-(m-tolyl)anilino]anthracene |
| 262°C | |
| C40H32N2 | |
| N,N′C-Diphenyl-N,N′C-di(m-tolyl)-9,10-anthracenediamine, N,N′C-Bis(3-methylphenyl)-N,N′C-diphenyl-9,10-anthracenediamine | |
| CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=CC=C6)C7=CC=CC(=C7)C | |
| 540.71 | |
| 540.71 | |
| Crystalline Powder |
Safety and Handling
TSCA : No