missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis[N-(m-tolyl)anilino]anthracene 98.0+%, TCI America™

Catalog No. B4965200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4965200MG 200 mg
B49651G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4965200MG Supplier TCI America Supplier No. B4965200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 189263-81-8
Molecular Formula C40H32N2
Molecular Weight (g/mol) 540.71
InChI Key LMTXHBDTVAIIQY-UHFFFAOYSA-N
Synonym N,N′C-Diphenyl-N,N′C-di(m-tolyl)-9,10-anthracenediamine, N,N′C-Bis(3-methylphenyl)-N,N′C-diphenyl-9,10-anthracenediamine
PubChem CID 22976649
IUPAC Name 9-N,10-N-bis(3-methylphenyl)-9-N,10-N-diphenylanthracene-9,10-diamine
SMILES CC1=CC(=CC=C1)N(C2=CC=CC=C2)C3=C4C=CC=CC4=C(C5=CC=CC=C53)N(C6=CC=CC=C6)C7=CC=CC(=C7)C

Specifications

Melting Point 262°C
Color Yellow
Quantity 200 mg
Formula Weight 540.71
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis[N-(m-tolyl)anilino]anthracene