missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis[N-(2-naphthyl)anilino]anthracene 98.0+%, TCI America™

Catalog No. p-7123904
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5023200MG 200 mg
B50231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5023200MG Supplier TCI America Supplier No. B5023200MG

Chemical Identifiers

CAS 473717-08-7
Molecular Formula C46H32N2
Molecular Weight (g/mol) 612.776
InChI Key LIBLRJBCMMHCHS-UHFFFAOYSA-N
Synonym N,N′-Di(naphthalen-2-yl)-N,N′-diphenylanthracene-9,10-diamine
PubChem CID 53404751
IUPAC Name 9-N,10-N-dinaphthalen-2-yl-9-N,10-N-diphenylanthracene-9,10-diamine
SMILES C1=CC=C(C=C1)N(C2=CC3=CC=CC=C3C=C2)C4=C5C=CC=CC5=C(C6=CC=CC=C64)N(C7=CC=CC=C7)C8=CC9=CC=CC=C9C=C8

Specifications

Quantity 200 mg
Formula Weight 612.78
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis[N-(2-naphthyl)anilino]anthracene