missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9,10-Bis(4-methoxyphenyl)anthracene 98.0+%, TCI America™
$192.00 - $559.22
Chemical Identifiers
| CAS | 24672-76-2 |
|---|---|
| Molecular Formula | C28H22O2 |
| Molecular Weight (g/mol) | 390.48 |
| MDL Number | MFCD00009936 |
| InChI Key | KTYCXBAOXVVIMM-UHFFFAOYSA-N |
| PubChem CID | 374567 |
| IUPAC Name | 9,10-bis(4-methoxyphenyl)anthracene |
| SMILES | COC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OC)C=C2)C2=CC=CC=C12 |
Chemical Identifiers
| 24672-76-2 | |
| 390.48 | |
| KTYCXBAOXVVIMM-UHFFFAOYSA-N | |
| 9,10-bis(4-methoxyphenyl)anthracene |
| C28H22O2 | |
| MFCD00009936 | |
| 374567 | |
| COC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OC)C=C2)C2=CC=CC=C12 |
Specifications
| 24672-76-2 | |
| C28H22O2 | |
| 200 mg | |
| COC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OC)C=C2)C2=CC=CC=C12 | |
| 390.48 | |
| 390.48 | |
| Crystalline Powder |
| 283°C | |
| MFCD00009936 | |
| KTYCXBAOXVVIMM-UHFFFAOYSA-N | |
| 9,10-bis(4-methoxyphenyl)anthracene | |
| 374567 | |
| ≥98.0% (GC) | |
| 9,10-Bis(4-methoxyphenyl)anthracene |
Safety and Handling
TSCA : No