missing translation for 'onlineSavingsMsg'
Learn More

9,10-Bis(4-methoxyphenyl)anthracene 98.0+%, TCI America™

Catalog No. B5022200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5022200MG 200 mg
B50221G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5022200MG Supplier TCI America Supplier No. B5022200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 24672-76-2
Molecular Formula C28H22O2
Molecular Weight (g/mol) 390.48
MDL Number MFCD00009936
InChI Key KTYCXBAOXVVIMM-UHFFFAOYSA-N
PubChem CID 374567
IUPAC Name 9,10-bis(4-methoxyphenyl)anthracene
SMILES COC1=CC=C(C=C1)C1=C2C=CC=CC2=C(C2=CC=C(OC)C=C2)C2=CC=CC=C12

Specifications

Melting Point 283°C
Quantity 200 mg
Formula Weight 390.48
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9,10-Bis(4-methoxyphenyl)anthracene