missing translation for 'onlineSavingsMsg'
Learn More

9,10-Anthraquinone, 98+%, Thermo Scientific Chemicals

Catalog No. p-7020691
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1273530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA1273530 Supplier Thermo Scientific Chemicals Supplier No. A1273530
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

Anthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Anthraquinone is used in paper industry as a catalyst to increase the pulp production yield and to improves the fiber strength through reduction reaction of cellulose to carboxylic acid. It is also used as a precursor for dye formation.

Solubility
Insoluble in water.

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.

Chemical Identifiers

CAS 84-65-1
Molecular Formula C14H8O2
Molecular Weight (g/mol) 208.22
MDL Number MFCD00001188
InChI Key RZVHIXYEVGDQDX-UHFFFAOYSA-N
Synonym anthraquinone, 9,10-anthraquinone, 9,10-anthracenedione, anthradione, hoelite, 9,10-dioxoanthracene, corbit, morkit, 9,10-anthrachinon, anthra-9,10-quinone
PubChem CID 6780
ChEBI CHEBI:40448
IUPAC Name anthracene-9,10-dione
SMILES O=C1C2=CC=CC=C2C(=O)C2=CC=CC=C12

Specifications

Melting Point 283°C to 287°C
Density 1.438
Boiling Point 377°C
Flash Point 185°C (365°F)
Odor Odorless
Quantity 250 g
Beilstein 390030
Merck Index 14,687
Solubility Information Insoluble in water.
Formula Weight 208.22
Percent Purity ≥98%
Chemical Name or Material 9,10-Anthraquinone
Show More Show Less
Hazard Category H317-H350
Hazard Statement GHS H Statement
H351-H317
Suspected of causing cancer.
May cause an allergic skin reaction.
Precautionary Statement P201-P202-P261-P272-P280g-P281-P302+P352-P308+P313-P333+P313-P363-P501c
EINECSNumber 201-549-0
RTECSNumber CB4725000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only