missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-(1-Naphthyl)carbazole 98.0+%, TCI America™
Supplier: TCI America N10125G
Specifications
| 9-(1-Naphthyl)carbazole | |
| 129°C | |
| C22H15N | |
| 5 g | |
| C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC2=CC=CC=C12 | |
| 293.37 | |
| 293.37 | |
| Crystalline Powder |
| 22034-43-1 | |
| White-Yellow | |
| MFCD26127424 | |
| QSOAYCUFEQGHDN-UHFFFAOYSA-N | |
| 9-(naphthalen-1-yl)-9H-carbazole | |
| 626218 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 22034-43-1 | |
| 293.37 | |
| QSOAYCUFEQGHDN-UHFFFAOYSA-N | |
| 9-(naphthalen-1-yl)-9H-carbazole |
| C22H15N | |
| MFCD26127424 | |
| 626218 | |
| C1=CC2=C(C=C1)C1=C(C=CC=C1)N2C1=CC=CC2=CC=CC=C12 |
Safety and Handling
TSCA : No