missing translation for 'onlineSavingsMsg'
Learn More

9-([1,1'-Biphenyl]-3-yl)-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7124128
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B55911G 1 g
B55915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B55911G Supplier TCI America Supplier No. B55911G
Only null left

Chemical Identifiers

CAS 1221237-87-1
Molecular Formula C24H17N
Molecular Weight (g/mol) 319.407
MDL Number MFCD28167065
InChI Key LKXFMLDAUIXMGY-UHFFFAOYSA-N
PubChem CID 58722663
IUPAC Name 9-(3-phenylphenyl)carbazole
SMILES C1=CC=C(C=C1)C2=CC(=CC=C2)N3C4=CC=CC=C4C5=CC=CC=C53

Specifications

Melting Point 128°C
Color White-Yellow
Quantity 1 g
Formula Weight 319.41
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-([1,1′-Biphenyl]-3-yl)-9H-carbazole