missing translation for 'onlineSavingsMsg'
Learn More
Learn More
9-([1,1'-Biphenyl]-3-yl)-9H-carbazole 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 128°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 319.41 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 9-([1,1′-Biphenyl]-3-yl)-9H-carbazole |
Safety and Handling
| TSCA | No |