missing translation for 'onlineSavingsMsg'
Learn More

9-([1,1'-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole 98.0+%, TCI America™

Catalog No. p-7123923
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5054200MG 200 mg
B50541G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5054200MG Supplier TCI America Supplier No. B5054200MG
Only null left

Chemical Identifiers

CAS 1533406-38-0
Molecular Formula C30H28BNO2
Molecular Weight (g/mol) 445.369
MDL Number MFCD28130384
InChI Key VQLXKJLKLNGTQF-UHFFFAOYSA-N
Synonym 9-(3-Biphenylyl)carbazole-3-boronic Acid Pinacol Ester, 2-[9-([1,1′C-Biphenyl]-3-yl)-9H-carbazol-3-yl]-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 90336757
IUPAC Name 9-(3-phenylphenyl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)carbazole
SMILES B1(OC(C(O1)(C)C)(C)C)C2=CC3=C(C=C2)N(C4=CC=CC=C43)C5=CC=CC(=C5)C6=CC=CC=C6

Specifications

Melting Point 238°C
Color White
Quantity 200 mg
Formula Weight 445.37
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 9-([1,1′-Biphenyl]-3-yl)-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole