missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ 850 Homogenizer Package

Description
- Includes 10 x 115mm, 20 x 195mm, and 30 x 195mm stainless steel generator probes
- Powerful 850 watt motor
- Integrated digital speed control displays speed and run time
- Variable speed control from 5,000 to 25,000rpm
- Can be used with plastic generator probes (adapter required for use)
- Processing range from 0.2mL to 5L
- Sound level: <72 db.
Certifications
CE
Warranty
Two years
Specifications
Specifications
| Speed | 5000 to 25,000 rpm |
| Voltage | 115 V |
| Electrical Requirements | 115 V, 50/60 Hz |
| Height (English) | 8.5 in. |
| Height (Metric) | 21.5 cm |
| For Use With (Application) | Tissue homogenization, tissue disruption, protein extraction, RNA extraction, DNA extraction, metabolite extraction, emulsification, proteomics, PCR, enzyme extraction, high speed stirring, powder suspension, genomics, metabolomics |
| Includes |
|
| Weight (English) | 56 oz. |
| Weight (Metric) | 1.6 kg |
| Product Type | Homogenizer |