missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Moldex™ 8000 Series Paint Spray/Pesticide Pre-Assembled Respirators
Description
- 8000 Series half-mask pre-assembled with OV cartridges, N95 pre-filters and pre-filter retainers
- For oil-free paint sprays and pesticides
- Soft, durable facepiece and lightweight design with extra-wide sealing
- Exhalation valve positioned for better communication
- Cartridges have a snap-in mount for easy replacement and no misthreading
- Living hinge allows head harness to be folded comfortably around neck when not in use
- Low profile, compact design fits well under welding helmets and with safety glasses
- Maintenance made simple with very few replacement parts
1 Includes two 19-003-346 N95 particulate prefilters, two 18-110 retainers, and two 18-102 organic vapor cartridges.
Specifications
Specifications
| Certifications (NIOSH) | NIOSH 42 CFR Part 84, TC-23C-1223 |
| Color | Gray |
| Filter Type | N95 |
| Material | Thermoplastic Elastomer |
| Product Type | Pre-Assembled Half-Mask Respirator |
| Size | Small |
| For Use With (Application) | Paint spray and pesticide applications which are oil-free |
| Includes | 8000 Facepiece, 8100 Organic Vapor Cartridges, 8910 N95 Pre-Filters and 8020 Pre-Filter Retainers |
| Model | 8111N |