Learn More
Hamilton™ 800 Microliter Syringes, No Needle Included
Attached to the flange of the 700 series syringe is a blue Syringe Holder that provides a spot to hold the syringe that will not heat up the barrel causing dispense inaccuracies. Additionally it provides support to small volume plungers that are prone to bending during injection. This series of syringes is ideal for dispensing volumes from 0.5 μL up to 250 μL.
Supplier:  Hamilton™ HAM764101
 View more versions of this product
Catalog No. 14824668
Only null left
Description
Description
  • Ideal for dispensing volumes from 0.5 μL up to 250 μL
  • A variety of different terminations and needle options are available
  • Syringe Holder is attached to the flange that prevents the barrel from heating up and provides support to small volume plungers that are prone to bending
Product Suggestions
Product Suggestions