missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Greiner Bio-One 8-well Polystyrene Round Bottom Strip Plates

Description
- 1 x 8 strips
- Non-Sterile
- Round Bottom
Specifications
Specifications
| Barcode | No Barcode |
| Format | Strip Plate |
| Array | 1 x 8 |
| Binding Type | Medium Binding |
| Color | Clear |
| Product Type | ELISA Strip Plate |
| Material | Polystyrene |
| No. of Wells | 8 |
| Sterility | Non-sterile |
| Surface Treatment | Medium Binding |
| Show More |