missing translation for 'onlineSavingsMsg'
Learn More

8-(Trifluoromethyl)quinoline, 98%, Thermo Scientific Chemicals

Catalog No. p-7053670
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6126703 1 g
AAH6126706 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6126703 Supplier Thermo Scientific Chemicals Supplier No. H6126703
Only null left

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 317-57-7
Molecular Formula C10H6F3N
Molecular Weight (g/mol) 197.16
MDL Number MFCD09744668
InChI Key AJXUSUNIYLSPER-UHFFFAOYSA-N
PubChem CID 12647568
IUPAC Name 8-(trifluoromethyl)quinoline
SMILES C1=CC2=C(C(=C1)C(F)(F)F)N=CC=C2

Specifications

Quantity 1 g
Synonym 8-trifluoromethyl quinoline, 8-cf3 quinoline, acmc-1ag4x, quinoline,8-trifluoromethyl, quinoline, 8-trifluoromethyl
Formula Weight 197.16
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 8-(Trifluoromethyl)quinoline
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only