Learn More
Applied Biosystems™ 8-strip Septa for SeqStudio™ Genetic Analyzer
Catalog No. A35643
Encompass
Change view
Click to view available options
Quantity:
24 septa
Catalog No. Quantity
A35643 24 septa
1 options
Catalog No. A35643 Supplier Applied Biosystems™ Supplier No. A35643
A radio-frequency identification tag is built into the frame for tracking critical run information, which is then reported on the software dashboard.
These are septa for 8-strip tubes for use on the SeqStudio genetic analyzer.
Order Info
24 Septa
Specifications
Product Line SeqStudio
Type Genetic Analyzer Septa
Quantity 24 septa
Shipping Condition Room Temperature
What does the SeqStudio Starter Kit (PN A35000) contain?
The SeqStudio Starter Kit (PN A35000) includes:
- SeqStudio Integrated Capillary Protector (Cat. No. A31923)
- SeqStudio Cathode Buffer Container (Cat. No. A33401)
- Sequencing Standard, BigDye Terminator v3.1, for 3500/SeqStudio Flex (Cat. No. 4404312)
- DS-33 GeneScan Installation Standards with GeneScan 600 LIZ Size Standard v2.0 (Cat. No. 4376911)
- Hi-Di Formamide (Cat. No. 4401457)
- MicroAmp Optical 96-Well Reaction Plate with Barcode (Cat. No. 4306737)
- MicroAmp 96-Well Tray/Retainer Set (Cat. No. 403081)
- MicroAmp Optical 8-Tube Strip, 0.2 mL (Cat. No. 4316567)
- Nuclease-Free Water (not DEPC-Treated) (Cat. No. AM9930)
- Cathode Buffer Container Reservoir Septa for SeqStudio Genetic Analyzer (Cat. No. A35640)
- 96-well Septa for SeqStudio Genetic Analyzer (Cat. No. A35641)
- 8-strip Septa for SeqStudio Genetic Analyzer (Cat. No. A35643)
- SeqStudio Cartridge v2 (PN A41328) (if this component is ordered online the catalog number is A41331)
How many times can I reuse the 8-strip Septa for SeqStudio Genetic Analyzer?
The 8-strip Septa for SeqStudio Genetic Analyzer are intended for single use only. We recommend replacing the strip septa after each run. If rinsed and reused, the septa can become sticky and gummy over time and may start sticking to the array. This may potentially increase the risk of damaging the array.
Bleach, detergents, or solvents should not be used to clean the septa as this can cause them to dry out faster and risk contaminating the system.
How often can the plate septa on the 3130/xl, 3730/xl, 3500/xl, and SeqStudio instruments be used?
We officially recommend that the plate septa be used one time only. However, we are aware that some of our customers will rinse the septa with water and re-use them. In those instances, the septa can become sticky and gummy over time and eventually start sticking to the array. In the best case scenario, the array will lift the plate off of the autosampler just enough to break the connection with the plate sensor and abort the run. The Event Log might show that the run aborted due to the user aborting it, so the run will appear to have completed or stopped with no errors and no data. The worst case scenario is that the plate remains on the array as the autosampler starts to move, bending the electrodes and breaking the array. We recommend replacing the plate septa regularly (replacing after each run is recommended). Do not use bleach, detergents, or solvents to clean the septa, it will cause them to dry out faster and also risks contaminating the system.
For Research Use Only. Not for use in diagnostic procedures.