missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinol Sulfate Monohydrate 98.0+%, TCI America™

Catalog No. Q001725G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q001725G 25 g
Q0017500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q001725G Supplier TCI America Supplier No. Q001725G

Chemical Identifiers

CAS 134-31-6
Molecular Formula C9H7NO
MDL Number MFCD00013095
Synonym Chinosol, 8-Hydroxyquinoline Sulfate, Oxine Sulfate, 8-Quinolinone Sulfate

Specifications

Melting Point 178°C
Color Yellow
Quantity 25 g
Formula Weight 388.39
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Quinolinol Sulfate Monohydrate
RTECSNumber VC8260000
TSCA Yes