Learn More
8-Quinolinesulfonyl chloride, 98%, Thermo Scientific™
$138.32 - $583.78
Chemical Identifiers
| CAS | 18704-37-5 |
|---|---|
| Molecular Formula | C9H6ClNO2S |
| Molecular Weight (g/mol) | 227.67 |
| InChI Key | JUYUYCIJACTHMK-UHFFFAOYSA-N |
| Synonym | 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp |
| PubChem CID | 29220 |
| IUPAC Name | quinoline-8-sulfonyl chloride |
| SMILES | C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
AC132210050
|
N/A | 5g |
Each for $138.32
|
|
|||||
|
AC132210250
|
N/A | 25g |
Each for $583.78
|
|
|||||
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 18704-37-5 | |
| 227.67 | |
| 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp | |
| quinoline-8-sulfonyl chloride |
| C9H6ClNO2S | |
| JUYUYCIJACTHMK-UHFFFAOYSA-N | |
| 29220 | |
| C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2 |
Specifications
| 18704-37-5 | |
| Authentic | |
| C9H6ClNO2S | |
| 8-quinolinesulfonyl chloride, 8-chlorosulfonyl-1-benzazine, 8-quinolylsulfonyl chloride, quinoline-8-sulfonic acid chloride, unii-5zik3tjs9m, 8-quinolinesulfonylchloride, quinoline-8-sulphonyl chloride, 5zik3tjs9m, pubchem5873, acmc-1busp | |
| JUYUYCIJACTHMK-UHFFFAOYSA-N | |
| quinoline-8-sulfonyl chloride | |
| 29220 | |
| 98% |
| 306.0°C | |
| 97.5% min. (Argentometry) | |
| 22,V,7,567 | |
| Solubility in water: reacts. | |
| C1=CC2=C(C(=C1)S(=O)(=O)Cl)N=CC=C2 | |
| 227.67 | |
| 227.67 | |
| 8-Quinolinesulfonyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: Rinse mouth. Do NOT induce vomiting. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower.
GHS Signal Word: Danger
EINECSNumber : 242-515-5
RUO – Research Use Only