missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinecarboxaldehyde 95.0+%, TCI America™

Catalog No. Q00961G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
Q00961G 1 g
Q00965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. Q00961G Supplier TCI America Supplier No. Q00961G

Chemical Identifiers

CAS 38707-70-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805837
InChI Key OVZQVGZERAFSPI-UHFFFAOYSA-N
Synonym 8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
PubChem CID 170103
IUPAC Name quinoline-8-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)N=CC=C2

Specifications

Melting Point 96°C
Color Yellow
Quantity 1 g
Formula Weight 157.17
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Quinolinecarboxaldehyde