missing translation for 'onlineSavingsMsg'
Learn More

8-Quinolinecarbaldehyde, Thermo Scientific™

Catalog No. p-2703609
Change view
Click to view available options
Quantity:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC04304DA 1g
CC04304DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC04304DA Supplier Thermo Scientific Chemicals Supplier No. CC04304DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 38707-70-9
Molecular Formula C10H7NO
Molecular Weight (g/mol) 157.172
MDL Number MFCD00805837
InChI Key OVZQVGZERAFSPI-UHFFFAOYSA-N
Synonym 8-quinolinecarboxaldehyde, 8-quinolinecarbaldehyde, quinoline-8-carboxaldehyde, 8-quinoline carbaldehyde, 8-formylquinoline, ccris 6968, timtec-bb sbb010129, 8-quinolinecarboaldehyde, acmc-209j1k, ksc224g8j
PubChem CID 170103
IUPAC Name quinoline-8-carbaldehyde
SMILES C1=CC2=C(C(=C1)C=O)N=CC=C2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1g
Formula Weight 157.17
Percent Purity 97%
Chemical Name or Material 8-Quinolinecarbaldehyde
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.