missing translation for 'onlineSavingsMsg'
Learn More

8-Pentadecanone 99.0+%, TCI America™

$223.68 - $652.41

Chemical Identifiers

CAS 818-23-5
Molecular Formula C15H30O
Molecular Weight (g/mol) 226.40
MDL Number MFCD00009555
InChI Key PQYGSSYFJIJDFK-UHFFFAOYSA-N
Synonym 8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
PubChem CID 13162
IUPAC Name pentadecan-8-one
SMILES CCCCCCCC(=O)CCCCCCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
D15455G
View Documents
TCI America
D15455G
5 g N/A
Only null left
 
D154525G
View Documents
TCI America
D154525G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

818-23-5
226.40
PQYGSSYFJIJDFK-UHFFFAOYSA-N
13162
CCCCCCCC(=O)CCCCCCC
C15H30O
MFCD00009555
8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
pentadecan-8-one

Specifications

818-23-5
White
C15H30O
5 g
PQYGSSYFJIJDFK-UHFFFAOYSA-N
pentadecan-8-one
13162
≥99.0% (GC)
8-Pentadecanone
43°C
140°C
MFCD00009555
8-pentadecanone, caprylone, heptyl ketone, diheptyl ketone, di-n-heptyl ketone, 8-oxopentadecane, pentadecane-8-one, saturated fatty acid, acmc-1bl8g, caprylone_gurudeebansatyavani
CCCCCCCC(=O)CCCCCCC
226.40
226.40
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

RTECSNumber : RZ2000000

TSCA : No

SDS
missing translation for 'documents'

missing translation for 'documents'