Learn More
8-Octanolactam, 98%
CAS: 935-30-8 | C8H15NO | 141.214 g/mol
$163.29 - $2040.70
Chemical Identifiers
| CAS | 935-30-8 |
|---|---|
| Molecular Formula | C8H15NO |
| Molecular Weight (g/mol) | 141.214 |
| MDL Number | MFCD00003273 |
| InChI Key | YDLSUFFXJYEVHW-UHFFFAOYSA-N |
| Synonym | 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo |
| PubChem CID | 13632 |
| IUPAC Name | azonan-2-one |
| SMILES | C1CCCC(=O)NCCC1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAA1862106
|
Thermo Scientific Chemicals
A1862106 |
5 g |
Each for $163.29
|
|
|||||
|
AAA1862114
|
Thermo Scientific Chemicals
A1862114 |
25 g |
Each for $612.56
|
|
|||||
|
AAA1862122
|
Thermo Scientific Chemicals
A1862122 |
100 g |
Each for $2,040.70
|
|
|||||
Description
2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Azacyclononanone has been used as an internal standard liquid chromatography-ultraviolet detection method for the determination of caprolactam in the aqueous food stimulant 3% acetic acid (w/v).
Solubility
Soluble in methanol (almost transparency).
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Chemical Identifiers
| 935-30-8 | |
| 141.214 | |
| YDLSUFFXJYEVHW-UHFFFAOYSA-N | |
| 13632 | |
| C1CCCC(=O)NCCC1 |
| C8H15NO | |
| MFCD00003273 | |
| 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo | |
| azonan-2-one |
Specifications
| 935-30-8 | |
| C8H15NO | |
| 5 g | |
| 2-azacyclononanone, caprylolactam, capryllactam, azacyclononan-2-one, 8-octanelactam, 8-aminooctanoic acid lactam, 2-perhydroazoninone, 2h-azonin-2-one, octahydro, cyclooctanone lactam, octamethylenimine, 2-oxo | |
| YDLSUFFXJYEVHW-UHFFFAOYSA-N | |
| azonan-2-one | |
| 13632 | |
| 98% |
| 72°C to 75°C | |
| MFCD00003273 | |
| 109775 | |
| Soluble in methanol (almost transparency). | |
| C1CCCC(=O)NCCC1 | |
| 141.214 | |
| 141.21 | |
| 8-Octanolactam |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 213-300-3
RTECSNumber : CO1757000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only