missing translation for 'onlineSavingsMsg'
Learn More

8-Methylquinoline, 97+%

Catalog No. AAL0328806
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0328806 5 g
AAL0328814 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0328806 Supplier Thermo Scientific Chemicals Supplier No. L0328806
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 611-32-5 | C10H9N | 143.19 g/mol

8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Methylquinoline has been used in preparation of osmium chloridophosphine complexes, as quinoline carbene tautomers.

Solubility
Soluble in water 0.1-0.5 g/100 mL at 17°C.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Chemical Identifiers

CAS 611-32-5
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006810
InChI Key JRLTTZUODKEYDH-UHFFFAOYSA-N
Synonym quinoline, 8-methyl, 8-methyl-quinoline, ccris 408, 8-methyl, o-toluquinoline, 8-metyl quinoline, 8-methyl quinoline, pubchem5806, acmc-209mpf, dsstox_cid_888
PubChem CID 11910
ChEBI CHEBI:48984
IUPAC Name 8-methylquinoline
SMILES CC1=C2N=CC=CC2=CC=C1

Specifications

Melting Point -80°C
Density 1.074
Boiling Point 134°C to 136°C (25 mmHg)
Flash Point 105°C (221°F)
Odor Unpleasant
Refractive Index 1.615
Quantity 5 g
Beilstein 111340
Solubility Information Soluble in water 0.1-0.5g/100 mL at 17°C.
Formula Weight 143.19
Percent Purity ≥97%
Chemical Name or Material 8-Methylquinoline
Show More Show Less
Hazard Category H302-H315-H319-H335-H341
Hazard Statement GHS H Statement
H341-H315-H319
Suspected of causing genetic defects.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P201-P202-P261-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
EINECSNumber 210-264-0
RTECSNumber VC0562000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only