missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-4-methylbenzo[g]coumarin 98.0+%, TCI America™

Catalog No. m1862100mg
Change view
Click to view available options
Quantity:
100 mg
Catalog No. Quantity
M1862100MG 100 mg
1 options

Catalog No. M1862100MG

Supplier: TCI America M1862100MG

Only null left

Chemical Identifiers

CAS 69169-71-7
Molecular Formula C15H12O3
Molecular Weight (g/mol) 240.26
MDL Number MFCD00445784
InChI Key GWFUYESISQUIHB-UHFFFAOYSA-N
Synonym 8-Methoxy-4-methyl-2H-naphtho[2,3-b]pyran-2-one
PubChem CID 12448345
IUPAC Name 8-methoxy-4-methyl-2H-benzo[g]chromen-2-one
SMILES COC1=CC=C2C=C3C(C)=CC(=O)OC3=CC2=C1

Specifications

Melting Point 221°C
Color Yellow
Quantity 100 mg
Formula Weight 240.26
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 8-Methoxy-4-methylbenzo[g]coumarin