missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxy-2-methylquinoline 98.0+%, TCI America™

Catalog No. M10881G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10881G 1 g
M10885G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10881G Supplier TCI America Supplier No. M10881G
Only null left

Chemical Identifiers

CAS 3033-80-5
Molecular Formula C11H11NO
Molecular Weight (g/mol) 173.215
MDL Number MFCD00034436
InChI Key OQXVXPBDSJQYRR-UHFFFAOYSA-N
Synonym 8-Methoxyquinaldine
PubChem CID 316986
IUPAC Name 8-methoxy-2-methylquinoline
SMILES CC1=NC2=C(C=CC=C2OC)C=C1

Specifications

Melting Point 124°C
Color White-Yellow
Quantity 1 g
Formula Weight 173.22
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Methoxy-2-methylquinoline