missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%

CAS: 207386-91-2 | C18H18N2O7S | 406.41 g/mol

$48.38 - $94.62

Chemical Identifiers

CAS 207386-91-2
Molecular Formula C18H18N2O7S
Molecular Weight (g/mol) 406.41
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0274314
View Documents
Thermo Scientific Chemicals
L0274314
25 g
Each for $48.38
Only null left
 
AAL0274322
View Documents
Thermo Scientific Chemicals
L0274322
100 g
Each for $94.62
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

207386-91-2
406.41
BNCXJZDIJIVJJO-UHFFFAOYSA-N
C18H18N2O7S
MFCD00149610
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

207386-91-2
C18H18N2O7S
25 g
8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard
406.41
99%
176°C to 179°C
MFCD00149610
3760550
BNCXJZDIJIVJJO-UHFFFAOYSA-N
406.42 (388.40 Anhydrous)
8-Hydroxyquinoline sulfate monohydrate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 205-137-1

RTECSNumber : VC8260000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only