missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline sulfate monohydrate, 98%

Catalog No. AAL0274314
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0274314 25 g
AAL0274322 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0274314 Supplier Thermo Scientific Chemicals Supplier No. L0274314

CAS: 207386-91-2 | C18H18N2O7S | 406.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 207386-91-2
Molecular Formula C18H18N2O7S
Molecular Weight (g/mol) 406.41
MDL Number MFCD00149610
InChI Key BNCXJZDIJIVJJO-UHFFFAOYSA-N
Synonym 8-quinolinol hemisulfate, 8-quinolinol hemisulfate hemihydrate, 8-hydroxyquinoline hemisulfate salt hemihydrate, quinolin-8-ol; sulfuric acid; hydrate, quinolin-8-ol sulfate hydrate 2:1:1, 8-hydroxyquinoline sulfate, 8-quinolinol sulfate 2:1 monohydrate, 8-quinolinol, sulfate, hydrate 2:1:1, 8-quinolinol hemisulfate salt hemihydrate, pestanal r , analytical standard

Specifications

Melting Point 176°C to 179°C
Quantity 25 g
Beilstein 3760550
Formula Weight 406.42 (388.40 Anhydrous)
Percent Purity 99%
Chemical Name or Material 8-Hydroxyquinoline sulfate monohydrate
Hazard Category H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P264b-P270-P301+P312-P330-P501c
EINECSNumber 205-137-1
RTECSNumber VC8260000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only