missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline Sulfate, 98%3, Spectrum™ Chemical

Catalog No. 18608293
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18608293 25 g Amber Glass Bottle
18608294 100 g Amber Glass Bottle
18608295 500 g Amber Glass Bottle
18608296 2.5 kg Amber Glass Bottle
18608297 12 kg Amber Glass Bottle
18608298 50 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18608293 Supplier Spectrum Chemical Mfg Cor Supplier No. H109625GM

Chemical Identifiers

CAS 134-31-6

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 25 g
Molecular Weight (g/mol) 388.4
Percent Purity 0.983
Grade Reagent
Physical Form Crystalline Powder or Lumps
Chemical Name or Material 8-Hydroxyquinoline Sulfate

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.