missing translation for 'onlineSavingsMsg'
Learn More
Learn More
8-Hydroxyquinoline N-Oxide 98.0+%, TCI America™
Supplier: TCI America H127225G
Specifications
| 8-Hydroxyquinoline N-Oxide | |
| 139°C | |
| MFCD00006739 | |
| 8-hydroxyquinoline-n-oxide, 8-hydroxyquinoline 1-oxide, 8-hydroxyquinoline n-oxide, 8-hydroxyquinolin-n-oxide, 8-quinolinol, 1-oxide, 8-quinolinol, n-oxide, quinolin-8-ol 1-oxide, o-hydroxyquinoline n-oxide, 8-hydroxyquinolin-1-ium-1-olate, quinoline-1,8-diol | |
| C1=CC2=C(C(=C1)O)[N+](=CC=C2)[O-] | |
| 161.16 | |
| 161.16 | |
| Crystalline Powder |
| 1127-45-3 | |
| C9H7NO2 | |
| 25 g | |
| FJKUOCCQEBLPNX-UHFFFAOYSA-N | |
| 1-oxidoquinolin-1-ium-8-ol | |
| 14312 | |
| ≥98.0% (HPLC,T) |
Chemical Identifiers
| 1127-45-3 | |
| 161.16 | |
| FJKUOCCQEBLPNX-UHFFFAOYSA-N | |
| 14312 | |
| C1=CC2=C(C(=C1)O)[N+](=CC=C2)[O-] |
| C9H7NO2 | |
| MFCD00006739 | |
| 8-hydroxyquinoline-n-oxide, 8-hydroxyquinoline 1-oxide, 8-hydroxyquinoline n-oxide, 8-hydroxyquinolin-n-oxide, 8-quinolinol, 1-oxide, 8-quinolinol, n-oxide, quinolin-8-ol 1-oxide, o-hydroxyquinoline n-oxide, 8-hydroxyquinolin-1-ium-1-olate, quinoline-1,8-diol | |
| 1-oxidoquinolin-1-ium-8-ol |
Safety and Handling
RTECSNumber : VC8238500
TSCA : No