missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic Acid Monohydrate 98.0+%, TCI America™

Catalog No. p-7130478
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H030625G 25 g
H0306500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H030625G Supplier TCI America Supplier No. H030625G
Only null left

Chemical Identifiers

CAS 283158-18-9
Molecular Formula C9H9NO5S
Molecular Weight (g/mol) 243.233
MDL Number MFCD00006795
InChI Key WUXYGIQVWKDVTJ-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
PubChem CID 2723649
IUPAC Name 8-hydroxyquinoline-5-sulfonic acid;hydrate
SMILES C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O

Specifications

Melting Point 314°C
Quantity 25 g
Formula Weight 225.22
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic Acid Monohydrate
RTECSNumber VC2570000
TSCA Yes