missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic acid monohydrate, 98%

Catalog No. p-4509660
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC121921000 100 g Plastic bottle
AC121925000 500 g Plastic bottle
AC121920025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC121921000 Supplier Thermo Scientific Chemicals Supplier No. 121921000

CAS: 283158-18-9 | C9H7NO4S·H2O | 243.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 283158-18-9
InChI Key WUXYGIQVWKDVTJ-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
PubChem CID 2723649
IUPAC Name 8-hydroxyquinoline-5-sulfonic acid;hydrate
SMILES C1=CC2=C(C=CC(=C2N=C1)O)S(=O)(=O)O.O

Specifications

Melting Point >300.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min
Packaging Plastic bottle
Molecular Formula C9H7NO4S·H2O
Quantity 100 g
Beilstein 22,407
Merck Index 14,4844
Solubility Information Solubility in water: freely soluble. Other solubilities: slightly soluble in most common organic solvents
Molecular Weight (g/mol) 243.24
Formula Weight 243.24
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic acid monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Suspected of causing genetic defects.
Precautionary Statement GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Use personal protective equipment as required.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lense

RUO – Research Use Only