missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic acid hydrate, 98%

Catalog No. p-7024804
Change view
Click to view available options
Quantity:
50 g
250 g
500 g
Catalog No. Quantity
AAB2002218 50 g
AAB2002230 250 g
AAB2002236 500 g
3 options

Catalog No. AAB2002218

Supplier: Thermo Scientific Chemicals B2002218

Only null left

CAS: 207386-92-3 | C9H7NO4S | 225.22 g/mol

It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a reagent for determination of trace metals. It is employed as pre-1,2 and post-column3 reagent for fluorescence detection of metal ions.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

Chemical Identifiers

CAS 207386-92-3
Molecular Formula C9H7NO4S
Molecular Weight (g/mol) 225.22
MDL Number MFCD00149482
InChI Key LGDFHDKSYGVKDC-UHFFFAOYSA-N
Synonym 8-hydroxyquinoline-5-sulfonic acid hydrate, 8-hydroxy-5-quinolinesulfonic acid monohydrate, 8-hydroxyquinoline-5-sulfonic acid monohydrate, unii-n85kc14z44, 8-hydroxy-5-quinolinesulfonic acid hydrate, 5-quinolinesulfonicacid, 8-hydroxy-, hydrate 1:1, 8-hydroxyquinoline-5-sulfonic acid, hydrate, acmc-209pxa, dsstox_cid_24317, dsstox_rid_80150
PubChem CID 2723649
IUPAC Name 8-hydroxyquinoline-5-sulfonic acid;hydrate
SMILES OC1=C2N=CC=CC2=C(C=C1)S(O)(=O)=O

Specifications

Melting Point 311°C to 313°C
Quantity 50 g
Beilstein 195247
Merck Index 14,4844
Solubility Information Soluble in water.
Formula Weight 225.22 (Anhydrous)
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline-5-sulfonic acid hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 201-570-5
RTECSNumber VC2570000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only