missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-2-carbonitrile, 98%

Catalog No. AAH5043303
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5043303 1 g
AAH5043306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH5043303 Supplier Thermo Scientific Chemicals Supplier No. H5043303
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6759-78-0 | C10H6N2O | 170.171 g/mol

8-Hydroxyquinoline-2-carbonitrile is a building block for preparing chelating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyquinoline-2-carbonitrile is a building block for preparing chelating agents.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Chemical Identifiers

CAS 6759-78-0
Molecular Formula C10H6N2O
Molecular Weight (g/mol) 170.171
MDL Number MFCD00216717
InChI Key KUQKKIBQVSFDHX-UHFFFAOYSA-N
PubChem CID 2734032
IUPAC Name 8-hydroxyquinoline-2-carbonitrile
SMILES C1=CC2=C(C(=C1)O)N=C(C=C2)C#N

Specifications

Melting Point 133°C to 136°C
Quantity 1 g
UN Number UN3439
Synonym 8-hydroxy-2-quinolinecarbonitrile, 8-hydroxyquinaldonitrile, 2-quinolinecarbonitrile,8-hydroxy, acmc-20alwc, 2-cyano-8-hydroxyquinoline, 8-hydroxy-2-quinolinecarbonitril, 2-quinolinecarbonitrile, 8-hydroxy, 8-hydroxy-2-quinolinecarbonitrile gc
Formula Weight 170.17
Percent Purity 98%
Chemical Name or Material 8-Hydroxyquinoline-2-carbonitrile
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H331-H302-H312-H315-H319-H335
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only