missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyjulolidine 97.0+%, TCI America™

Catalog No. H09285G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H09285G 5 g
H09281G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H09285G Supplier TCI America Supplier No. H09285G

Chemical Identifiers

CAS 41175-50-2
Molecular Formula C12H15NO
Molecular Weight (g/mol) 189.258
MDL Number MFCD00006918
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
PubChem CID 170474
SMILES C1CC2=C3C(=C(C=C2)O)CCCN3C1

Specifications

Melting Point 132°C
Quantity 5 g
Formula Weight 189.26
Percent Purity ≥97.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Hydroxyjulolidine