missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxy-2-methylquinoline, 98%

Catalog No. AAL0698936
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0698936 500 g
AAL0698922 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0698936 Supplier Thermo Scientific Chemicals Supplier No. L0698936
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 826-81-3 | C10H9NO | 159.188 g/mol

8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxy-2-methylquinoline is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 826-81-3
Molecular Formula C10H9NO
Molecular Weight (g/mol) 159.188
MDL Number MFCD00006765
InChI Key NBYLBWHHTUWMER-UHFFFAOYSA-N
Synonym 8-hydroxyquinaldine, 2-methyl-8-hydroxyquinoline, 2-methyl-8-quinolinol, 8-quinolinol, 2-methyl, 2-methyloxine, 8-hydroxy-2-methylquinoline, hydroxyquinaldine, 8-hydroxyqinaldine, 2-methyl-quinolin-8-ol, 2-methyl 8-quinolonol
PubChem CID 13224
IUPAC Name 2-methylquinolin-8-ol
SMILES CC1=NC2=C(C=CC=C2O)C=C1

Specifications

Melting Point 70°C to 74°C
Boiling Point 267°C
Quantity 500 g
UN Number UN3077
Beilstein 119194
Solubility Information Insoluble in water.
Formula Weight 159.19
Percent Purity 98%
Chemical Name or Material 8-Hydroxy-2-methylquinoline
DOTInformation Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber 212-562-6
RTECSNumber VC7920000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only