missing translation for 'onlineSavingsMsg'
Learn More

[8-(Glycidyloxy)-n-octyl]trimethoxysilane 97.0+%, TCI America™

Catalog No. p-7130277
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
G04695G 5 g
G046925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. G04695G Supplier TCI America Supplier No. G04695G
Only null left

Chemical Identifiers

CAS 1239602-38-0
Molecular Formula C14H30O5Si
Molecular Weight (g/mol) 306.474
InChI Key QYJYJTDXBIYRHH-UHFFFAOYSA-N
PubChem CID 102529357
IUPAC Name trimethoxy-[8-(oxiran-2-ylmethoxy)octyl]silane
SMILES CO[Si](CCCCCCCCOCC1CO1)(OC)OC

Specifications

Color Colorless
Quantity 5 g
Formula Weight 306.47
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material [8-(Glycidyloxy)-n-octyl]trimethoxysilane