missing translation for 'onlineSavingsMsg'
Learn More

Labconco 8 ft. Nexus Horizontal Clean Bench

Nexus Horizontal Clean Benches are designed to protect non-hazardous applications by providing an ISO Class 4 clean aairflow environment. The large, 5 in. touchscreen interface features Nexus OS™ software that simplifies operation, maintains records and provides user customizaton of the system.

Supplier:  Labconco 5381100

 View more versions of this product

Catalog No. 17195021

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

  • 8 ft. Nexus Horizontal Clean Bench
  • Internal work area
  • ISO Class 4 laminar aairflow is 10x cleaner than the industry standard ISO Class 5
  • Energy efficient ECM motor with CAP™ Technology ensures accurate aairflow between certifications
Specifications

Chemical Identifiers

Horizontal Clean Bench
HEPA
50.1 in.
127.5 cm
None
Epoxy
Optic White
30.2 in.
76.7 cm
NEMA 5-20P

Specifications

Horizontal Clean Bench
HEPA
Stainless Steel
26 in.
50.1 in.
127.5 cm
Clean Bench Only, LED
NEMA 5-20P
101.0 in.
256.6 cm
Epoxy
LED
Optic White
66.6 cm
30.2 in.
76.7 cm
None
115 V
98.6 in.
250.4 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions