missing translation for 'onlineSavingsMsg'
Learn More

8-Dimethylamino-1-octanol 93.0+%, TCI America™

Catalog No. D28465G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D28465G 5 g
D28461G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D28465G Supplier TCI America Supplier No. D28465G
Only null left

Chemical Identifiers

CAS 29823-87-8
Molecular Formula C10H23NO
Molecular Weight (g/mol) 173.3
MDL Number MFCD03701100
InChI Key CAKJEDBOIMYCHP-UHFFFAOYSA-N
PubChem CID 9815358
IUPAC Name 8-(dimethylamino)octan-1-ol
SMILES CN(C)CCCCCCCCO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 173.30
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Dimethylamino-1-octanol