missing translation for 'onlineSavingsMsg'
Learn More

8-Chlorotheophylline 98.0+%, TCI America™

Catalog No. C029325G
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C029325G 25 g
C0293250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C029325G Supplier TCI America Supplier No. C029325G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 85-18-7
Molecular Formula C7H7ClN4O2
Molecular Weight (g/mol) 214.61
MDL Number MFCD00005581
InChI Key RYIGNEOBDRVTHA-UHFFFAOYSA-N
Synonym 8-chlorotheophylline, 1,3-dimethyl-8-chloroxanthine, theophylline, 8-chloro, 8-chloro-theophylline, unii-ge2ua340fm, 8-chloro-1,3-dimethyl-1h-purine-2,6 3h,7h-dione, 1h-purine-2,6-dione, 8-chloro-3,7-dihydro-1,3-dimethyl, 8-chloro-3,7-dihydro-1,3-dimethyl-1h-purine-2,6-dione, ge2ua340fm, chembl88611
PubChem CID 10661
ChEBI CHEBI:59771
IUPAC Name 8-chloro-1,3-dimethyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
SMILES CN1C2=C(NC(Cl)=N2)C(=O)N(C)C1=O

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 214.61
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 8-Chlorotheophylline
EINECSNumber (9)-0379
RTECSNumber XH5063000
TSCA No