missing translation for 'onlineSavingsMsg'
Learn More

8-Chloroquinoline 98.0+%, TCI America™

$259.90 - $916.38

Chemical Identifiers

CAS 611-33-6
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00047618
InChI Key RUSMDMDNFUYZTM-UHFFFAOYSA-N
Synonym quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
PubChem CID 69139
ChEBI CHEBI:48985
IUPAC Name 8-chloroquinoline
SMILES ClC1=C2N=CC=CC2=CC=C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C02845G
View Documents
TCI America
C02845G
5 g N/A
Only null left
 
C028425G
View Documents
TCI America
C028425G
25 g N/A
Only null left
 
Description

Description

Specifications

Chemical Identifiers

611-33-6
163.60
RUSMDMDNFUYZTM-UHFFFAOYSA-N
69139
8-chloroquinoline
C9H6ClN
MFCD00047618
quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
CHEBI:48985
ClC1=C2N=CC=CC2=CC=C1

Specifications

611-33-6
288°C
MFCD00047618
quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
ClC1=C2N=CC=CC2=CC=C1
163.60
CHEBI:48985
≥98.0% (GC)
8-Chloroquinoline
Yellow
C9H6ClN
5 g
RUSMDMDNFUYZTM-UHFFFAOYSA-N
8-chloroquinoline
69139
163.60
Liquid
Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents