missing translation for 'onlineSavingsMsg'
Learn More

8-Chloroquinoline, 99%

Catalog No. AC405020250
Change view
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC405020250 25 g Glass bottle
AC405020100 10 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC405020250 Supplier Thermo Scientific Chemicals Supplier No. 405020250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 611-33-6 | C9H6ClN | 163.60 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 611-33-6
Molecular Formula C9H6ClN
Molecular Weight (g/mol) 163.60
MDL Number MFCD00047618
InChI Key RUSMDMDNFUYZTM-UHFFFAOYSA-N
Synonym quinoline, 8-chloro, ccris 3982, 8-choloroquinoline, 8-chloro-quinoline, pubchem10742, acmc-1aumj, ksc163s9l, 8-chloro-quinolin;quinoline, 8-chloro
PubChem CID 69139
ChEBI CHEBI:48985
IUPAC Name 8-chloroquinoline
SMILES ClC1=C2N=CC=CC2=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -20°C
Color Yellow to Brown
Boiling Point 288.5°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Solubility Information Solubility in water: soluble
Formula Weight 163.61
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 8-Chloroquinoline, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 210-265-6
TSCA TSCA

RUO – Research Use Only