missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-2-methylquinoline, 98%

Catalog No. aab2050403
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2050403 1 g
AAB2050406 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2050403 Supplier Thermo Scientific Chemicals Supplier No. B2050403
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3033-82-7 | C10H8ClN | 177.631 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3033-82-7
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00051764
InChI Key VVLYDFPOGMTMFJ-UHFFFAOYSA-N
Synonym 8-chloroquinaldine, quinaldine, 8-chloro, quinoline, 8-chloro-2-methyl, 8-chloro-2-methyl-quinoline, 2-methyl-8-chloroquinoline, pubchem5852, acmc-1aehk, maybridge1_006766, 8-chloranyl-2-methyl-quinoline, 8-chloro-2-methylquinoline
PubChem CID 221113
IUPAC Name 8-chloro-2-methylquinoline
SMILES CC1=NC2=C(C=CC=C2Cl)C=C1

Specifications

Melting Point 65°C to 68°C
Quantity 1 g
Beilstein 119193
Formula Weight 177.63
Percent Purity 98%
Chemical Name or Material 8-Chloro-2-methylquinoline
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only