missing translation for 'onlineSavingsMsg'
Learn More

8-Chloro-1-n-octanol 97.0+%, TCI America™

Catalog No. C24485ML
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C24485ML 5 mL
C244825ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C24485ML Supplier TCI America Supplier No. C24485ML

Chemical Identifiers

CAS 23144-52-7
Molecular Formula C8H17ClO
Molecular Weight (g/mol) 164.673
MDL Number MFCD00040005
InChI Key YDFAJMDFCCJZSI-UHFFFAOYSA-N
Synonym 8-chloro-1-octanol, 1-octanol, 8-chloro, 8-chlorooctanol, 8-chloro-octan-1-ol, 8-chloro-1-n-octanol, 8-chlor-1-octanol, 8-chlor-octan-1-ol, acmc-1cr4x
PubChem CID 90012
IUPAC Name 8-chlorooctan-1-ol
SMILES C(CCCCCl)CCCO

Specifications

Color Yellow
Boiling Point 130°C
Quantity 5 mL
Formula Weight 164.67
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 8-Chloro-1-n-octanol