Learn More
Applied Biosystems™ 3500 Genetic Analyzer 8-Capillary Array, 36 cm
Description
Order Info
Each
Specifications
Specifications
| For Use With (Application) | Human Identification, Sequencing, Fragment Analysis |
| Quantity | 1 array |
| For Use With (Equipment) | 3500 Genetic Analyzer |
| Length (Metric) | 36 cm (Capillary) |
| Product Type | Genetic Analyzer 8-Capillary Array |
| Capillary Length | 36 cm |
| Shipping Condition | Room Temperature |
| Content And Storage | Store this array at ambient temperature. For details about storing the capillary array on and off the instrument, please refer to the Applied Biosystems 3500/3500xL Genetic Analyzer User Guide. |
| Description | 3500 Genetic Analyzer 8-Capillary Array, 36cm Capillary, Internally Uncoated Capillaries, For Use With 3500 Genetic Analyzer, 15 to 30°C Storage Temperature |
Frequently Asked Questions (FAQs)
Beginning with 3500 Data Collection Software version 3, the 3500 Data Collection Software does not support automatic secondary analysis. This is also indicated in the software release notes. To perform secondary data analysis, you will need to manually import the .fsa file into the GeneMapper software. For optimal performance, we recommend that you install the GeneMapper Software on a stand-alone computer that is not part of the 3500 Genetic Analyzer system.
If the array is stuck in the pump, even after loosening the knob, squirt around the edge of the array head with distilled water and try to remove it again. If the array will not move, a service call may be required. Do not use excessive force or tools to try and remove it.
Some of the other causes of an Unstable Electrophoresis current detected error message are:
1.Leak on the system
2. Polymer that:
-Has expired
-Has been left on the instrument for more than the recommended time
-Is a mixture of expired polymer and non-expired polymer
3. Running Buffer that was:
-On the instrument longer than 2 weeks
-Not filled to the fill line or evaporated below the fill line
4. An arcing event that was not cleaned afterwards using the water wash wizard
5. Not performing regular maintenance on the instrument
6. Hardware issues
Inspect the system for leaks. If you do not see any leaks on the system, perform the wash the pump chamber and channels wizard using the conditioning pouch and place fresh, non-expired polymer and fresh Anode and Cathode buffer containers. If the problem persists, a service call may be required.
If we do not specify the expiry date or number of uses, the warranty will last for 12 months from the date of shipment.
The Applied Biosystems 3500/3500xl Genetic Analyzer capillary arrays are warrantied for a specific number of runs or until the expiration date printed on the array package, whichever comes first. If you notice that you are starting to get reduced data quality (e.g., shorter reads, trailing peaks in all 4 colors) even with the Sequencing Standard, these may be signs that the array requires changing.
For Research Use Only. Not for use in diagnostic procedures.