missing translation for 'onlineSavingsMsg'
Learn More
Learn More
National Safety Apparel™ Enespro™ 8 Cal Ultrasoft Khaki Coverall
Durable and comfortable ultrasoft coveralls offer a unique blend of 88% cotton and 12% high-tenacity nylon for a smooth feel. The inherently FR material delivers exceptional performance and reliability.
Supplier: National Safety Apparel™ C88UKLG32
Description
- Action back allows for full range of motion
- All metal covered to ensure no contact with skin
- Chest pockets with flaps and hidden snap closures
- Comfortable sewn-in elastic waist for better fit
- Convenient side pass-through pockets for access to pant pockets
- Covered zipper front for easy entry
- FR fabric will not melt, drip or ignite
- FR protection from electrical arc flash and flash fire hazards
- Home laundry
- Leg openings made to fit over work boots
- Two front pockets with pass-through for access to pants pockets
Specifications
| Coveralls | |
| Unisex | |
| Khaki | |
| Elastic (Stitched-in) | |
| Collared | |
| Zipper | |
| Non-sterile | |
| Flame | |
| NFPA 70E, NFPA 2112, CSA Z462, ASTM F1506, OSHA 1910.269 | |
| Utilities, Oil and Gas, Electrical Contractors |
| Large | |
| Cotton, Nylon | |
| Reusable | |
| Open | |
| Unhooded | |
| Stitched | |
| CAT 2 | |
| 8 cal/cm² | |
| None | |
| Coveralls |